Please login to your online account to display your discounted pricing

anti-CDX2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human CDX2 /CDX3 was used as the immunogen for the antibody.
Antigen CDX2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224583

 
novus biologicals canada
NBP224583
Unlabeled 0.1mg Each for $257.41

NBP224583SS

 
novus biologicals canada
NBP224583SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human CDX2 /CDX3 was used as the immunogen for the antibody.
Antigen CDX2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the GATA-4 antibody is 100% homologous in human, 93% homologous in rhesus monkey, 87% homologous in rat, 82% in mouse, and 81% homologous in dog.
Gene Accession No. Q99626
Gene ID (Entrez) 1045
Formulation 100μg of antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL