Please login to your online account to display your discounted pricing

anti-Cdk4 Polyclonal, Thermo Scientific™

Cdk4 Polyclonal antibody PA1-12580 from Thermo Scientific™ specifically detects Cdk4 in Human samples, and it is validated for Immunoprecipitation and Western Blot

$448.05 - $448.05

Specifications

Antigen Cdk4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen TrpE-fusion protein containing the C-terminal 164 residues of human Cdk4.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-801

 
pierce biotechnology inc
PA112580
Unlabeled 100μg Each for $448.05
Description & Specifications

Specifications

Antigen Cdk4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen TrpE-fusion protein containing the C-terminal 164 residues of human Cdk4.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Cdk4 Polyclonal antibody PA1-12580 from Thermo Scientific™ specifically detects Cdk4 in Human samples, and it is validated for Immunoprecipitation and Western Blot

Cdk4 is involved in the control of the cell cycle.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: TrpE-fusion protein containing the C-terminal 164 residues of human Cdk4.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: CMM3, MGC14458, PSK-J3
Uniprot Accession: P11802
Gene Symbol: CDK4
Entrez GeneID: 1019
PA1-12580 detects Cdk4 in Human samples. PA1-12580 has been successfully used in Immunoprecipitation and Western Blot procedures. PA1-12580 The immunogen is TrpE-fusion protein containing the C-terminal 164 residues of human Cdk4..