Please login to your online account to display your discounted pricing

anti-CD97 Polyclonal, Thermo Scientific™


CD97 Polyclonal Ab PA5-33413 from Thermo Scientific™ specifically detects CD97 in Hu samples, and it is validated for ICC and IHC(P)

Manufacturer: pierce biotechnology inc  PA533413

 View more versions of this product

Catalog No. PIPA533413

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene ID (Entrez) 976
Applications Immunocytochemistry
Applications Immunohistochemistry (Paraffin)
Antigen CD97
Gene Accession No. P48960
Gene Alias CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen
Gene Symbols CD97
Host Species Rabbit
Species Reactivity Human
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Format Purified
Cross Reactivity Human
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Isotype IgG
Quantity 50μg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled

CD97 Polyclonal Ab PA5-33413 from Thermo Scientific™ specifically detects CD97 in Hu samples, and it is validated for ICC and IHC(P)

CD97 is a 75-85kDa monomeric glycoprotein that is present on the surface of most activated leucocytes. It is a receptor involved in both cell adhesion and signaling. CD97 is a member of the G protein coupled receptor family. It is expressed abundantly in cells of hematopoietic origin and is markedly upregulated on activated T and B cells. ESTs have been isolated from a wide variety of tissue libraries.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen
Uniprot Accession: P48960
Entrez GeneID: 976
Gene Symbol: CD97
Percent identity with other species by BLAST analysis: Human (100%) Monkey (95%).