Please login to your online account to display your discounted pricing

anti-CD97 Polyclonal, Thermo Scientific™

CD97 Polyclonal Ab PA5-33413 from Thermo Scientific™ specifically detects CD97 in Hu samples, and it is validated for ICC and IHC(P)

$506.05 - $506.05

Specifications

Antigen CD97
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533414

 
pierce biotechnology inc
PA533414
Unlabeled 50μg Each for $506.05

PIPA533413

 
pierce biotechnology inc
PA533413
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen CD97
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

CD97 Polyclonal Ab PA5-33413 from Thermo Scientific™ specifically detects CD97 in Hu samples, and it is validated for ICC and IHC(P)

The Vasoactive Intestinal Polypeptide Receptor 2 (VPAC2) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from N-terminal extracellular domain of human CD97.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen
Uniprot Accession: P48960
Gene Symbol: CD97
Entrez GeneID: 976
PA5-34306 detects VPAC2/VIPR2 in Human samples. PA5-34306 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-34306 The immunogen is Synthetic 18 amino acid peptide from C-terminus of human VIPR2..