anti-CD90.2, FITC, Clone: 30-H12, EMD Millipore™
Rat Monoclonal Antibody
Manufacturer:  Millipore Sigma MABF1448
 View more versions of this product
Catalog No. MABF1448MI
Description
Description
Specifically detects CD90.2 Clone: 30-H12 in Mouse samples, and it is validated for Flow Cytometry
The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.
Specifications
Specifications
CD90.2
30-H12
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, pH 7.2.
Purified
Rat
IgG2b
25μg
Inflammation, Immunology
Primary
Murine
Flow Cytometry
FITC
P01831
Thy1, Thy-1
Proprietary immunogen
Purified by size-exclusion chromatography
RUO
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Monoclonal