missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CD8a Red Fluor 710, Clone: 53-6.7, EMD Millipore™

Rat Monoclonal Antibody

Manufacturer:  Millipore Sigma MABF1553

 View more versions of this product

Catalog No. MABF1553MI



Description

Description

Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry

The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.
Specifications

Specifications

CD8a
53-6.7
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Purified
Rat
Purified by size-exclusion chromatography
RUO
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Monoclonal
Flow Cytometry
Red Fluor 710
P01731
CD8a
Proprietary immunogen
100μg
Inflammation, Immunology
Primary
Murine
Documents

For Research Use Only