missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CD8, Clone: 53-6.7, R&D Systems

$138.08 - $138.08

Specifications

Antigen CD8
Clone 53-6.7
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MLDP4 R & D Systems
RDSMLDP4
1 Vial Each for $138.08
Specifications

Description

Applications:

Cell Depletion

Description & Specifications

Specifications

CD8
Affinity Purified
RUO
IgG2a
Primary
53-6.7
Rat
Mouse thymus or spleen.
Monoclonal