Please login to your online account to display your discounted pricing

anti-CD8, Clone: 53-6.7, R&D Systems


Manufacturer: r & d systems  RDSMLDP4

 View more versions of this product

Catalog No. MLDP4

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Rat
Clone 53-6.7
Isotype IgG2a
Antigen CD8
Immunogen Mouse thymus or spleen.
Quantity 1 Vial
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified