Please login to your online account to display your discounted pricing

anti-CD5, Clone: 53-7.3, R&D Systems


Manufacturer: r & d systems  RDSMAB115

 View more versions of this product

Catalog No. MAB115

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Format Purified
Host Species Rat
Immunogen Mouse thymus or spleen.
Isotype IgG2a
Quantity 500μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 53-7.3
Conjugate Unlabeled
Cross Reactivity Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant

CD5 Monoclonal antibody MAB115 Specifically detects CD5 in Mouse samples. It is validated for Blocking (B Cell Proliferation), Cytotoxicity Assay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation