Please login to your online account to display your discounted pricing

anti-CD5, Clone: 53-7.3, R&D Systems

$139.08 - $432.57

Specifications

Antigen CD5
Clone 53-7.3
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB115

 
r & d systems
RDSMAB115
Unlabeled 500μg Each for $432.57

MLDP3

 
r & d systems
RDSMLDP3
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen CD5
Clone 53-7.3
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse thymus or spleen.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion