Please login to your online account to display your discounted pricing

anti-CD5, Clone: 53-7.3, R&D Systems


Manufacturer: r & d systems  RDSMLDP3

 View more versions of this product

Catalog No. MLDP3

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Clone 53-7.3
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen Mouse thymus or spleen.
Isotype IgG2a
Quantity 1 Vial
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Murine