missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CD45.2 Red Fluor 710, Clone: 104, EMD Millipore™

Mouse Monoclonal Antibody

Manufacturer:  Millipore Sigma MABF1476

 View more versions of this product

Catalog No. MABF1476MI



Description

Description

Specifically detects CD45.2 Clone: 104 in Mouse samples, and it is validated for Flow Cytometry

The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.
Specifications

Specifications

CD45.2
104
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Purified
Murine
Purified by size-exclusion chromatography
RUO
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Monoclonal
Flow Cytometry
Red Fluor 710
P06800
Ptprc, Ly-5
Proprietary immunogen
100μg
Inflammation, Immunology
Primary
Murine
Documents

For Research Use Only