Please login to your online account to display your discounted pricing

anti-CD4, Clone: GK1.5, R&D Systems

$139.08 - $139.08

Specifications

Antigen CD4
Clone GK1.5
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MLDP2

 
r & d systems
RDSMLDP2
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen CD4
Clone GK1.5
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion