Please login to your online account to display your discounted pricing

anti-CD4, Clone: GK1.5, R&D Systems


Manufacturer: r & d systems  RDSMLDP2

 View more versions of this product

Catalog No. MLDP2

  • / Each
 

Description & Specifications

Specifications

Antigen CD4
Isotype IgG2b
Regulatory Status RUO
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Quantity 1 Vial
Primary or Secondary Primary
Conjugate Unlabeled
Clone GK1.5
Monoclonal or Polyclonal Monoclonal