Please login to your online account to display your discounted pricing

anti-CD36/SR-B3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227414

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227414

  • / Each
 

Description & Specifications

Specifications

Antigen CD36/SR-B3
Applications Western Blot
Concentration 0.3mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 150μg in 500μL TBS with 50% glycerol, 0.5mg/mL BSA.
Host Species Rabbit
Immunogen Human CD36 amino acids 99-124.
Quantity 0.15mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 948
Target Species Human