Please login to your online account to display your discounted pricing

anti-CD36/SR-B3, Polyclonal, Novus Biologicals™

$142.00 - $409.37

Specifications

Host Species Rabbit
Immunogen Human CD36 amino acids 99-124.
Antigen CD36/SR-B3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227414

 
novus biologicals canada
NBP227414
Unlabeled 0.15mg Each for $409.37

NBP227414SS

 
novus biologicals canada
NBP227414SS
Unlabeled 0.025mg Each for $142.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Human CD36 amino acids 99-124.
Antigen CD36/SR-B3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 948
Formulation 150μg in 500μL TBS with 50% glycerol, 0.5mg/mL BSA.
Concentration 0.3mg/mL