Please login to your online account to display your discounted pricing

anti-CD36/SR-B3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227414SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227414SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Conjugate Unlabeled
Concentration 0.3mg/mL
Formulation 150μg in 500μL TBS with 50% glycerol, 0.5mg/mL BSA.
Gene ID (Entrez) 948
Quantity 0.025mg
Antigen CD36/SR-B3
Applications Western Blotting
Host Species Rabbit
Format Affinity Purified
Cross Reactivity Human
Primary or Secondary Primary
Immunogen Human CD36 amino acids 99-124.