Please login to your online account to display your discounted pricing

CD36 Polyclonal Antibody, Invitrogen™


CD36 Antibody, PA1-16814, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: Western Blot.

Manufacturer: pierce biotechnology inc  PA116814

Catalog No. APPA6814

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL purified antibody (1mg/mL) in PBS and 0.02% sodium azide
Gene ID (Entrez) 948
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Applications Western Blotting
Gene Accession No. P16671
Gene Alias FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Species Reactivity Human
Storage Requirements Store at 4° C
Antigen CD36
Gene Symbols CD36
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region of human CD36 between residues 300-400.
Quantity 100μL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled

CD36 Antibody, PA1-16814, from Invitrogen. Species Reactivity: Human, Mouse; Applications: Western Blot.

CD36 is a cell surface class B scavenger receptor. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signalling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. May function as a cell adhesion molecule. Directly mediates cyto adherence of Plasmodium falciparum parasitized erythrocytes. Binds long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Isotype: IgG
Immunogen: A synthetic peptide mapping to a region of human CD36 between residues 300-400.
Formulation: 100μL purified antibody (1mg/mL) in PBS and 0.02% sodium azide
Synonyms: FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Uniprot Accession: P16671
Entrez GeneID: 948
Gene Symbol: CD36
The target sequence has 95% sequence homology with rat, monkey, Golden hamster and rabbit, 90% sequence homology with mouse, and 75% sequence homology with bovine.