Please login to your online account to display your discounted pricing

anti-CD36 Polyclonal, Thermo Scientific™

CD36 Polyclonal antibody PA1-16815 from Thermo Scientific™ specifically detects CD36 in Human samples, and it is validated for Western Blot

$506.05 - $506.05

Specifications

Antigen CD36
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

APPA6815

 
pierce biotechnology inc
PA116815
Biotin 100μL Each for $506.05
Description & Specifications

Specifications

Antigen CD36
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region of human CD36 between residues 300-400.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 4° C

CD36 Polyclonal antibody PA1-16815 from Thermo Scientific™ specifically detects CD36 in Human samples, and it is validated for Western Blot

CD36 is a cell surface class B scavenger receptor. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signalling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. May function as a cell adhesion molecule. Directly mediates cyto adherence of Plasmodium falciparum parasitized erythrocytes. Binds long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: A synthetic peptide mapping to a region of human CD36 between residues 300-400.
Formulation: 100μL purified Antibody (0.9mg/mL) in PBS, with no preservative;
Synonyms: FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Uniprot Accession: P16671
Gene Symbol: CD36
Entrez GeneID: 948
PA1-16815 detects CD36 in Human samples. PA1-16815 has been successfully used in Western Blot procedures. PA1-16815 The immunogen is A synthetic peptide mapping to a region of human CD36 between residues 300-400..