missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human CD31/PECAM1 (within residues 100-300). [Swiss-Prot P16284]
Antigen CD31/PECAM-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP171663SS Novus Biologicals Canada
NBP171663SS
0.025mL Each for $139.00
NBP171663 Novus Biologicals Canada
NBP171663
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
CD31/PECAM-1
Affinity Purified
RUO
P16284
PBS, 30% glycerol
A genomic peptide made to an internal region of human CD31/PECAM1 (within residues 100-300). [Swiss-Prot P16284]
Polyclonal
Primary
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
5175
0.6mg/mL