Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Fluorescein, Clone: 9G11, R&D Systems


Manufacturer: r & d systems  FAB3567F100

 View more versions of this product

Catalog No. FAB3567F100

  • / Each
 

Description & Specifications

Specifications

Antigen CD31/PECAM-1
Applications Flow Cytometry
Clone 9G11
Conjugate Fluorescein
Cross Reactivity Human
Format Purified
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Host Species Murine
Immunogen Activated HUVEC human umbilical vein endothelial cells
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Test Specificity Detects human CD31/PECAM‑1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E‑Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed.
Target Species Human