Please login to your online account to display your discounted pricing

anti-CD3, Clone: 17A2, R&D Systems

$142.08 - $142.08

Specifications

Antigen CD3
Clone 17A2
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MLDP1

 
r & d systems
RDSMLDP1
Unlabeled 1 Vial Each for $142.08
Description & Specifications

Specifications

Antigen CD3
Clone 17A2
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Immunogen T cell hybridoma D1.
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion