Please login to your online account to display your discounted pricing

anti-CD3, Clone: 17A2, R&D Systems


Manufacturer: r & d systems  RDSMLDP1

 View more versions of this product

Catalog No. MLDP1

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Regulatory Status RUO
Cross Reactivity Mouse
Antigen CD3
Quantity 1 Vial
Immunogen T cell hybridoma D1.
Isotype IgG2b
Clone 17A2
Format Affinity Purified
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal