missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD23, PE-Cyanine7, clone: B3B4, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS25023282

 View more versions of this product

Catalog No. 501129372



Description

Description

CD23 is a 45kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Specifications

Specifications

CD23
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P20693
CD23, CD23A, CLEC4J, FCE2, IGEBF, BLAST-2, C-type lectin domain family 4, member J, CD23 antigen, fc-epsilon-RII, immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, lymphocyte IgE receptor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
B3B4
PE-Cyanine7
Fcer2a
Liquid
Fcer2a
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14128
Documents

For Research Use Only.