Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056337SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056337T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity In MCF-7, a 42 kDa band should be observed.
Concentration 0.5mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 1234
Gene Accession No. ABN10015.1
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Format Purified
Host Species Rabbit
Applications Simple Western
Applications Western Blotting
Cross Reactivity Human
Immunogen A synthetic peptide corresponding to amino acids within 300 and 350 of human CCR5 was used as immunogen.
Quantity 0.025mg
Regulatory Status RUO
Antigen CCR5