Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056337

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056337

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Gene Accession No. ABN10015.1
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1234
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Test Specificity In MCF-7, a 42 kDa band should be observed.
Concentration 0.5mg/mL
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Antigen CCR5
Quantity 0.1mg
Immunogen A synthetic peptide corresponding to amino acids within 300 and 350 of human CCR5 was used as immunogen.
Cross Reactivity Human