Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100531SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100531SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Gene ID (Entrez) 1234
Format Affinity Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications In vitro Assay
Applications Western Blotting
Test Specificity This antibody is specific for the linear epitope detected by the monoclonal antibody 2D7. Human and rhesus.
Concentration 0.5mg/mL
Host Species Rabbit
Quantity 0.025mL
Conjugate Unlabeled
Antigen CCR5
Cross Reactivity Human
Cross Reactivity Primate
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide demonstrating a high affinity towards the monoclonal CCR5 2D7. This peptide is known as the 2D7-2SK peptide and consists of the following residues: EWQKEGLVTLWLGGGGSC.
Regulatory Status RUO