Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of the human CCR5 protein (between amino acids 1-350) [UniProt P51681]
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231374SS

 
novus biologicals canada
NBP231374SS
Unlabeled 0.025mg Each for $109.00

NBP231374

 
novus biologicals canada
NBP231374
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of the human CCR5 protein (between amino acids 1-350) [UniProt P51681]
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Immunogen has 86% homology to bovine.
Gene Accession No. P51681
Gene ID (Entrez) 1234
Formulation PBS
Concentration 1.0mg/mL