Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids within 300 and 350 of human CCR5 was used as immunogen.
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056337

 
novus biologicals canada
NB10056337
Unlabeled 0.1mg Each for $255.80

NB10056337T

 
novus biologicals canada
NB10056337SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids within 300 and 350 of human CCR5 was used as immunogen.
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Test Specificity In MCF-7, a 42 kDa band should be observed.
Gene Accession No. ABN10015.1
Gene ID (Entrez) 1234
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL