Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™

$139.00 - $139.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide demonstrating a high affinity towards the monoclonal CCR5 2D7. This peptide is known as the 2D7-2SK peptide and consists of the following residues: EWQKEGLVTLWLGGGGSC.
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100531SS

 
novus biologicals canada
NB100531SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide demonstrating a high affinity towards the monoclonal CCR5 2D7. This peptide is known as the 2D7-2SK peptide and consists of the following residues: EWQKEGLVTLWLGGGGSC.
Antigen CCR5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications In vitro Assay
Applications Western Blot
Test Specificity This antibody is specific for the linear epitope detected by the monoclonal antibody 2D7. Human and rhesus.
Gene ID (Entrez) 1234
Formulation PBS
Concentration 0.5mg/mL