Please login to your online account to display your discounted pricing

anti-CCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100531

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100531

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 1234
Concentration 0.5mg/mL
Test Specificity This antibody is specific for the linear epitope detected by the monoclonal antibody 2D7. Human and rhesus.
Format Affinity Purified
Quantity 0.1mL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications In vitro Assay
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Antigen CCR5
Primary or Secondary Primary
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Immunogen A synthetic peptide demonstrating a high affinity towards the monoclonal CCR5 2D7. This peptide is known as the 2D7-2SK peptide and consists of the following residues: EWQKEGLVTLWLGGGGSC.