Please login to your online account to display your discounted pricing

anti-CCR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP148337SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP148337SS

  • / Each
 

Description & Specifications

Specifications

Antigen CCR2
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P41597
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human CCR2 protein (within residues 20-100). [Swiss-Prot No. P00338]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 729230
Test Specificity Human. Reactivity with Rat reported by varified customer. The immunogen sequence similarity with other species: Rhesus monkey and several other primate species (100%), Mouse (69%).
Target Species Human
Target Species Rat