Please login to your online account to display your discounted pricing

anti-CCR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP148337

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP148337

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene ID (Entrez) 729230
Test Specificity Human. Reactivity with Rat reported by varified customer. The immunogen sequence similarity with other species: Rhesus monkey and several other primate species (100%), Mouse (69%).
Concentration 1mg/mL
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. P41597
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human CCR2 protein (within residues 20-100). [Swiss-Prot No. P00338]
Regulatory Status RUO
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Rat
Antigen CCR2
Monoclonal or Polyclonal Polyclonal