Please login to your online account to display your discounted pricing

anti-CCR1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide of human CCR1 protein.
Antigen CCR1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056334T

 
novus biologicals canada
NB10056334SS
Unlabeled 0.025mg Each for $139.00

NB10056334

 
novus biologicals canada
NB10056334
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide of human CCR1 protein.
Antigen CCR1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In A375, a 42 kDa band should be observed.
Gene Accession No. NP_001286.1
Gene ID (Entrez) 1230
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL