Please login to your online account to display your discounted pricing

anti-CCR1, Alexa Fluor 488, Clone: 53504, R&D Systems

$458.10 - $458.10

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P32246
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

FAB145G100

 
r & d systems
FAB145G100
100 Tests Each for $458.10
Description & Specifications

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P32246

Host Species: Mouse
Clone: 53504
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: Swiss 3T3 mouse cell line transfected with human CCR1.

Applications:

Flow Cytometry