Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107034SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107034SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Human and Rat. Mouse reactivity reported by a customer review. Immunogen sequence has 95% homology to mouse.
Conjugate Unlabeled
Antigen CCL2/MCP1
Formulation PBS, 30% glycerol
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot No. P14844]
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Concentration 1.42mg/mL
Format Affinity Purified
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Rat
Host Species Rabbit