Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107034

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107034

  • / Each
 

Description & Specifications

Specifications

Antigen CCL2/MCP1
Applications Western Blot
Concentration 1.42mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot No. P14844]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6347
Test Specificity Human and Rat. Mouse reactivity reported by a customer review. Immunogen sequence has 95% homology to mouse.
Target Species Human
Target Species Rat