missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot No. P14844]
Antigen CCL2/MCP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP107034 Novus Biologicals Canada
NBP107034
0.1mL Each for $309.05
NBP107034SS Novus Biologicals Canada
NBP107034SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
CCL2/MCP1
Affinity Purified
RUO
Human and Rat. Mouse reactivity reported by a customer review. Immunogen sequence has 95% homology to mouse.
PBS, 30% glycerol
Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot No. P14844]
Polyclonal
Primary
Western Blot
6347
1.42mg/mL