Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107035SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107035SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Applications Blocking
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Neutralization
Applications Western Blotting
Gene ID (Entrez) 6347
Regulatory Status RUO
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Concentration 1.22mg/mL
Cross Reactivity Human
Cross Reactivity Rat
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 15-40). [Swiss-Prot No. P14844]
Quantity 0.025mL
Primary or Secondary Primary
Format Affinity Purified
Host Species Rabbit
Antigen CCL2/MCP1