Please login to your online account to display your discounted pricing

anti-Cav1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142820

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142820

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Antigen Cav1.2
Test Specificity Human, mouse, rat, rabbit and other vertebrates.
Conjugate Unlabeled
Gene Accession No. Q13936
Gene ID (Entrez) 775
Formulation PBS
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Host Species Rabbit
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Primary or Secondary Primary
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of rat CACNA1C (within residues 1150-1200). [Swiss-Prot No. P22002]