Please login to your online account to display your discounted pricing

anti-Cav1.2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat CACNA1C (within residues 1150-1200). [Swiss-Prot No. P22002]
Antigen Cav1.2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142820

 
novus biologicals canada
NBP142820
Unlabeled 0.1mL Each for $329.00

NBP142820SS

 
novus biologicals canada
NBP142820SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat CACNA1C (within residues 1150-1200). [Swiss-Prot No. P22002]
Antigen Cav1.2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blot
Test Specificity Human, mouse, rat, rabbit and other vertebrates.
Gene Accession No. Q13936
Gene ID (Entrez) 775
Formulation PBS
Concentration 1mg/mL