Please login to your online account to display your discounted pricing

anti-Cav1.2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142820SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142820SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene Accession No. Q13936
Immunogen Synthetic peptide made to an internal portion of rat CACNA1C (within residues 1150-1200). [Swiss-Prot No. P22002]
Conjugate Unlabeled
Gene ID (Entrez) 775
Concentration 1mg/mL
Primary or Secondary Primary
Test Specificity Human, mouse, rat, rabbit and other vertebrates.
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO
Antigen Cav1.2
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal