Please login to your online account to display your discounted pricing

anti-Catalase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224916

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224916

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 847
Gene Accession No. P04040
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Immunogen The catalase enzyme from bovine liver was used as the immunogen.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Quantity 0.1mg
Conjugate Unlabeled
Host Species Rabbit
Antigen Catalase