Please login to your online account to display your discounted pricing

anti-Catalase, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen The catalase enzyme from bovine liver was used as the immunogen.
Antigen Catalase
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224916

 
novus biologicals canada
NBP224916
Unlabeled 0.1mg Each for $307.00

NBP224916SS

 
novus biologicals canada
NBP224916SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The catalase enzyme from bovine liver was used as the immunogen.
Antigen Catalase
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P04040
Gene ID (Entrez) 847
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL