Please login to your online account to display your discounted pricing

anti-Catalase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224916SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224916SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. P04040
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Regulatory Status RUO
Gene ID (Entrez) 847
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Catalase
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen The catalase enzyme from bovine liver was used as the immunogen.
Format Purified
Quantity 0.025mg
Primary or Secondary Primary