Please login to your online account to display your discounted pricing

anti-Caspase-9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056120SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056120T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This polyclonal antisera recognizes the proform of caspase-9 (∽50 kDa), and the large (∽35 kDa) subunit (with pro-domain sequences) of active/cleaved Caspase-9. It does not recognize the small (∽15 kDa) subunit of active/cleaved Caspase-9. Intermediate caspase-9 cleavage forms may also be seen at ∽21 kDa.
Gene ID (Entrez) 842
Formulation PBS
Format Antiserum
Cross Reactivity Human
Immunogen A synthetic peptide corresponding to amino acids 113-130 (PEIRKPEVLRPETPRPVD) of human caspase-9 was used as immunogen; GenBank no. gi|55960247|emb|CAI12974.1|.
Antigen Caspase-9
Quantity 0.025mL
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit