Please login to your online account to display your discounted pricing

anti-Caspase-9, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to amino acids 188-206 (EKLRRRFSS*LHFMVEVKGD) of human caspase-9 was used as immunogen; GenBank no. gi|27802691|gb|AAO21133.1|. S* refers to phosphorylated serine.
Antigen Caspase-9
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056122T

 
novus biologicals canada
NB10056122SS
Unlabeled 0.025mL Each for $144.63

NB10056122

 
novus biologicals canada
NB10056122
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to amino acids 188-206 (EKLRRRFSS*LHFMVEVKGD) of human caspase-9 was used as immunogen; GenBank no. gi|27802691|gb|AAO21133.1|. S* refers to phosphorylated serine.
Antigen Caspase-9
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Caspase-9 .
Gene ID (Entrez) 842
Formulation PBS