Please login to your online account to display your discounted pricing

anti-Caspase-9, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 113-130 (PEIRKPEVLRPETPRPVD) of human caspase-9 was used as immunogen; GenBank no. gi|55960247|emb|CAI12974.1|.
Antigen Caspase-9
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056120T

 
novus biologicals canada
NB10056120SS
Unlabeled 0.025mL Each for $144.63

NB10056120

 
novus biologicals canada
NB10056120
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 113-130 (PEIRKPEVLRPETPRPVD) of human caspase-9 was used as immunogen; GenBank no. gi|55960247|emb|CAI12974.1|.
Antigen Caspase-9
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This polyclonal antisera recognizes the proform of caspase-9 (∽50 kDa), and the large (∽35 kDa) subunit (with pro-domain sequences) of active/cleaved Caspase-9. It does not recognize the small (∽15 kDa) subunit of active/cleaved Caspase-9. Intermediate caspase-9 cleavage forms may also be seen at ∽21 kDa.
Gene ID (Entrez) 842
Formulation PBS