Please login to your online account to display your discounted pricing

anti-Caspase 9 (Carboxy-Terminal Divergent), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056366

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056366

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene ID (Entrez) 842
Concentration 0.5mg/mL
Test Specificity A 45 kDa band is detected.
Applications Western Blotting
Immunogen This antibody was raised against a mixture of two synthetic peptides within amino acids 328-383 of rat Caspase-9 CTD.
Host Species Rabbit
Regulatory Status RUO
Antigen Caspase 9 (Carboxy-Terminal Divergent)
Quantity 0.2mL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Format Purified