Please login to your online account to display your discounted pricing

anti-Caspase 9 (Carboxy-Terminal Divergent), Polyclonal, Novus Biologicals™

$139.00 - $274.59

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a mixture of two synthetic peptides within amino acids 328-383 of rat Caspase-9 CTD.
Antigen Caspase 9 (Carboxy-Terminal Divergent)
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056366

 
novus biologicals canada
NB10056366
Unlabeled 0.2mL Each for $274.59

NB10056366T

 
novus biologicals canada
NB10056366SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a mixture of two synthetic peptides within amino acids 328-383 of rat Caspase-9 CTD.
Antigen Caspase 9 (Carboxy-Terminal Divergent)
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity A 45 kDa band is detected.
Gene ID (Entrez) 842
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL